7-Chinolincarbaldehyd 95%

Lieferant: Apollo Scientific
Achtung

OR924120-100MG OR924120-250MG
APOSOR924120-100MGEA 0 CHF
APOSOR924120-100MG APOSOR924120-250MG
7-Chinolincarbaldehyd 95%
7-Chinolincarbaldehyd
Formel: C₁₀H₇NO
Lager Temperatur: Raumtemperatur
MDL Number: MFCD01366121
CAS-Nummer: 49573-30-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR