1,1,3,3-Tetramethyldisiloxan

Lieferant: Apollo Scientific

APOSOR70109-250GEA 254 CHF
APOSOR70109-250G
1,1,3,3-Tetramethyldisiloxan
1,1,3,3-Tetramethyldisiloxan
Formel: C₄H₁₄OSi₂
MW: 134,33 g/mol
Siedepunkt: 70…71 °C
Dichte: 0,76 g/cm³ (25 °C)
Flash-Pt: –26 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00008256
CAS-Nummer: 3277-26-7
UN: 1993
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR