1-Bromisochinolin

Lieferant: Apollo Scientific
Achtung

APOSOR59454-1GEA 114 CHF
APOSOR59454-1G
1-Bromisochinolin
1-Bromoisoquinoline
Formel: C₉H₆BrN
MW: 208,0546 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00234478
CAS-Nummer: 1532-71-4
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR