Methyl-4-aminobenzoat 95%

Lieferant: Apollo Scientific
Achtung

Synonyms: Methyl 4-aminobenzoate

OR55419-1KG
APOSOR55419-1KGEA 0 CHF
APOSOR55419-1KG
Methyl-4-aminobenzoat 95%
Methyl-4-aminobenzoat
Formel: C₈H₉NO₂
MW: 151,16 g/mol
Melting Pt: 110…114 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00007891
CAS-Nummer: 619-45-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR