6-Bromchinazolin
Lieferant: Apollo Scientific
|
Achtung
|
OR54428-5G
APOSOR54428-5GEA
133
CHF
APOSOR54428-5G
6-Bromchinazolin
6-Bromoquinazoline
Formel:
C₈H₅BrN₂ MW: 209,0400 g/mol Melting Pt: 304.2 °C Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD08236695 CAS-Nummer: 89892-21-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...