N-Benzhydryl-N-methylamine

Lieferant: Apollo Scientific
Gefahr

Synonyms: N-Methylbenzhydrylamine

OR5228-1G OR5228-500MG
APOSOR5228-1GEA 0 CHF
APOSOR5228-1G APOSOR5228-500MG
N-Benzhydryl-N-methylamine
N-Benzhydryl-N-methylamine
Formel: C₁₄H₁₅N
Melting Pt: 38…42 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00467853
CAS-Nummer: 14683-47-7
UN: 3259
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR