N-Fmoc-D-valin 99%

Lieferant: Apollo Scientific

APOSOR52196-100GEA 133 CHF
APOSOR52196-100G
N-Fmoc-D-valin 99%
N-Fmoc-D-valin
Formel: C₂₀H₂₁NO₄
MW: 339,39 g/mol
Melting Pt: 143…144 °C
Lager Temperatur: Kühlschrank
MDL Number: MFCD00062953
CAS-Nummer: 84624-17-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR