N-Boc-D-valin 99%
Lieferant: Apollo Scientific
APOSOR51864-100GEA
116
CHF
APOSOR51864-100G
N-Boc-D-valin 99%
N-Boc-D-valin
Formel:
C₁₀H₁₉NO₄ MW: 217,3 g/mol Melting Pt: 76…80 °C Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00038282 CAS-Nummer: 22838-58-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...