Isocytosin 99%
Lieferant: Apollo Scientific
APOSOR480710-100GEA
717
CHF
APOSOR480710-100G
Isocytosin 99%
Isocytosine
Formel:
C₄H₅N₃O MW: 111,10 g/mol Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00057557 CAS-Nummer: 108-53-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...