N-(tert-Butoxycarbonyl)-D-2-phenylglycine 98%

Lieferant: Apollo Scientific

APOSOR480572-100GEA 90.8 CHF
APOSOR480572-100G
N-(tert-Butoxycarbonyl)-D-2-phenylglycine 98%
N-(tert-Butoxycarbonyl)-D-2-phenylglycine
Formel: C₁₃H₁₇NO₄
MW: 251,28 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00062043
CAS-Nummer: 33125-05-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR