D-Propargylglycine 98%

Lieferant: Apollo Scientific

APOSOR480427-5GEA 201 CHF
APOSOR480427-5G
D-Propargylglycine 98%
D-Propargylglycine
Formel: C₅H₇NO₂
MW: 113,116 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00237407
CAS-Nummer: 23235-03-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR