Tetrahydro-2H-thiopyran-4-amin-1,1-dioxidhydrochlorid 97%
Lieferant: Apollo Scientific
Achtung
Synonyms: (1,1-Dioxidotetrahydro-2H-thiopyran-4-yl)amine Hydrochloride, 4-Aminotetrahydro-2H-thiopyran 1,1-dioxide hydrochloride hydrochloride, (1,1-Dioxidotetrahydro-2H-thiopyran-4-yl)amine hydrochloride
OR46016-1G OR46016-5G OR46016-25G
APOSOR46016-1GEA 83.6 CHF
APOSOR46016-1G APOSOR46016-5G APOSOR46016-25G
Tetrahydro-2H-thiopyran-4-amin-1,1-dioxidhydrochlorid 97%
4-Aminotetrahydro-2H-thiopyran 1,1-dioxide hydrochloride
Formel: C₅H₁₂ClNO₂S
MW: 185,6700 g/mol
Melting Pt: 288…289 °C
Lager Temperatur: Kühlschrank
MDL Number: MFCD11114559
CAS-Nummer: 116529-31-8