1-Brombutan
Lieferant: Apollo Scientific
APOSOR460056-100GEA
102
CHF
APOSOR460056-100G
1-Brombutan
1-Brombutan
Formel:
BrH₂C(CH₂)₂CH₃ MW: 137,02 g/mol Siedepunkt: 100…104 °C Melting Pt: –112 °C Dichte: 1,276 g/cm³ (25 °C) Flash-Pt: 10 °C Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00000260 CAS-Nummer: 109-65-9 UN: 1126 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...