4-(4-Bromobenzyl)morpholine
Lieferant: Apollo Scientific
|
Gefahr
|
OR3763-1G
OR3763-5G
APOSOR3763-1GEA
26.2
CHF
APOSOR3763-1G
APOSOR3763-5G
4-(4-Bromobenzyl)morpholine
4-(4-Bromobenzyl)morpholine
Formel:
C₁₁H₁₄BrNO Melting Pt: 85 °C Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00979535 CAS-Nummer: 132833-51-3 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...