2-(Cyanmethoxy)phenylboronsäure 98%

Lieferant: Apollo Scientific
Achtung

OR361023-250MG OR361023-100MG
APOSOR361023-250MGEA 0 CHF
APOSOR361023-250MG APOSOR361023-100MG
2-(Cyanmethoxy)phenylboronsäure 98%
2-(Cyanmethoxy)phenylboronsäure
Formel: C₈H₈BNO₃
Lager Temperatur: Kühlschrank
MDL Number: MFCD09908171
CAS-Nummer: 947533-29-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR