2,3-Pyridindicarbonsäure

Lieferant: Apollo Scientific
Achtung

APOSOR315023-100GEA 64.6 CHF
APOSOR315023-100G
2,3-Pyridindicarbonsäure
2,3-Pyridindicarbonsäure
Formel: C₇H₅NO₄
MW: 167,12 g/mol
Melting Pt: 189…190 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00006295
CAS-Nummer: 89-00-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR