4-Amino-7-methoxy-2-methylchinolin

Lieferant: Apollo Scientific

APOSOR307953-1GEA 542 CHF
APOSOR307953-1G
4-Amino-7-methoxy-2-methylchinolin
4-Amino-7-methoxy-2-methylquinoline
Formel: C₁₁H₁₂N₂O
Lager Temperatur: Raumtemperatur
MDL Number: MFCD09787723
CAS-Nummer: 66735-41-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR