3-Amino-2,5-dichlorpyridin 96%
Lieferant: Apollo Scientific
|
Achtung
|
APOSOR29881-5GEA
28
CHF
APOSOR29881-5G
APOSOR29881-10G
APOSOR29881-1G
3-Amino-2,5-dichlorpyridin 96%
3-Amino-2,5-dichlorpyridin
Formel:
C₅H₄Cl₂N₂ MW: 163,01 g/mol Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00204157 CAS-Nummer: 78607-32-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...