2-Brom-6-methoxypyridin
Lieferant: Apollo Scientific
|
Achtung
|
APOSOR2983-5GEA
28.6
CHF
APOSOR2983-5G
APOSOR2983-1G
APOSOR2983-25G
2-Brom-6-methoxypyridin
2-Bromo-6-methoxypyridine
Formel:
C₆H₆BrNO MW: 188,02 g/mol Siedepunkt: 206 °C Dichte: 1,53 g/cm³ (25 °C) Flash-Pt: 104 °C Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00088345 CAS-Nummer: 40473-07-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...