6-Chlor-m-toluidin 97%
Lieferant: Apollo Scientific
|
Achtung
|
APOSOR2643-1GEA
17.6
CHF
APOSOR2643-1G
APOSOR2643-25G
APOSOR2643-5G
6-Chlor-m-toluidin 97%
6-Chlor-m-toluidin
Formel:
C₇H₈ClN MW: 141,6 g/mol Siedepunkt: 228…230 °C Melting Pt: 29…30 °C Flash-Pt: 107 °C Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00007674 CAS-Nummer: 95-81-8 EINECS: 202-454-7 UN: 2239 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...