Ferrocenmethanol 97%
Lieferant: Apollo Scientific
APOSOR24509-5GEA
99.4
CHF
APOSOR24509-5G
Ferrocenmethanol 97%
Ferrocenmethanol
Formel:
C₁₁H₁₂FeO MW: 216,06 g/mol Melting Pt: 79…81 °C Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00040878 CAS-Nummer: 1273-86-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...