4'-Isobutylacetophenon 97%

Lieferant: Apollo Scientific
Achtung

Synonyms: p-Isobutylacetophenone

OR22079-250G OR22079-50G
APOSOR22079-250GEA 375 CHF
APOSOR22079-250G APOSOR22079-50G
4'-Isobutylacetophenon 97%
4'-Isobutylacetophenon
Formel: C₁₂H₁₆O
MW: 176,26 g/mol
Siedepunkt: 107 °C/2mmHg
Dichte: 0.952 g/cm³ (25 °C)
Flash-Pt: 40 °C
Lager Temperatur: Kühlschrank
MDL Number: MFCD00027393
CAS-Nummer: 38861-78-8
EINECS: 254-159-8
UN: 1224
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR