Fmoc-L-3,3-diphenylalanine 97%

Lieferant: Apollo Scientific

APOSOR14628-1GEA 105 CHF
APOSOR14628-1G
Fmoc-L-3,3-diphenylalanine 97%
Fmoc-L-3,3-diphenylalanine
Formel: C₃₀H₂₅NO₄
MW: 463,533 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00672337
CAS-Nummer: 201484-50-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR