Methyl-2-chloro-6-methylpyrimidine-4-carboxylate

Lieferant: Apollo Scientific
Achtung

OR1236-1G OR1236-5G
APOSOR1236-1GEA 0 CHF
APOSOR1236-1G APOSOR1236-5G
Methyl-2-chloro-6-methylpyrimidine-4-carboxylate
Methyl-2-chloro-6-methylpyrimidine-4-carboxylate
Formel: C₇H₇ClN₂O₂
Melting Pt: 108…113 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00117906
CAS-Nummer: 89793-11-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR