Methyltricosanoat

Lieferant: Apollo Scientific

Synonyms: Tricosansäuremethylester

APOSOR111025-1GEA 392 CHF
APOSOR111025-1G
Methyltricosanoat
Methyltricosanoat
Formel: C₂₄H₄₈O₂
MW: 368,64 g/mol
Melting Pt: 53…56 °C
Lager Temperatur: Kühlschrank
MDL Number: MFCD00009349
CAS-Nummer: 2433-97-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR