6-Iod-1H-indazol
Lieferant: Apollo Scientific
|
Achtung
|
OR110288-100G
OR110288-25G
APOSOR110288-100GEA
0
CHF
APOSOR110288-100G
APOSOR110288-25G
6-Iod-1H-indazol
6-Iodo-1H-indazole
Formel:
C₇H₅IN₂ Melting Pt: 208…211 °C Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD04114695 CAS-Nummer: 261953-36-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...