Thidiazuron
Lieferant: Apollo Scientific
|
Achtung
|
BIT0888-5G
BIT0888-1G
BIT0888-25G
APOSBIT0888-5GEA
166
CHF
APOSBIT0888-5G
APOSBIT0888-1G
APOSBIT0888-25G
Thidiazuron
Thidiazuron
Compound with cytokin like activity.
Formel:
C₉H₈N₄OS MW: 220.25 g/mol Melting Pt: 210.5…212.5 °C Lager Temperatur: Gefrierschrank |
MDL Number:
MFCD00078723 CAS-Nummer: 51707-55-2 EINECS: 257-356-7 UN: 3077 ADR: 9+EHS III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...