GMBS (N-Succinimidyl 4-maleimidobutyrate)

Lieferant: Apollo Scientific

Synonyms: GMBS

APOSBICL226-100MGEA 0 CHF
APOSBICL226-100MG
GMBS (N-Succinimidyl 4-maleimidobutyrate)
GMBS (N-Succinimidyl 4-maleimidobutyrate)

Primary amine reactive -Sulphhydryl reactive. Protein conjugation Hapten carrier Protein conjugation

Formel: C₁₂H₁₂N₂O₆
MW: 280,24 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00036817
CAS-Nummer: 80307-12-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR