[(tert-Butoxycarbonyl)aminooxy]acetic acid ≥98%
Lieferant: Apollo Scientific
APOSBIBP1015-5GEA
227
CHF
APOSBIBP1015-5G
[(tert-Butoxycarbonyl)aminooxy]acetic acid ≥98%
[(tert-Butoxycarbonyl)aminooxy]acetic acid
Formel:
C₇H₁₃NO₅ MW: 191,18 g/mol Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD01632027 CAS-Nummer: 42989-85-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...