Bilirubin

Lieferant: Apollo Scientific

APOSBIB0134-1GEA 201 CHF
APOSBIB0134-1G
Bilirubin
Bilirubin
Formel: C₃₃H₃₆N₄O₆
MW: 584,67 g/mol
Melting Pt: 192 °C
Lager Temperatur: Gefrierschrank
MDL Number: MFCD00005499
CAS-Nummer: 635-65-4
EINECS: 211-239-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR