2-(Cyclohexylamino)ethansulfonsäure (CHES)

Lieferant: Apollo Scientific
Achtung

APOSBI3458-250GEA 0 CHF
APOSBI3458-250G APOSBI3458-100G APOSBI3458-500G
2-(Cyclohexylamino)ethansulfonsäure (CHES)
2-(Cyclohexylamino)ethansulfonsäure (CHES)

  • Assay (by titration) 98+%
  • Water (by Karl Fischer) <1.0%
  • Biological buffer - useful range 8.6-10.0

Formel: C₈H₁₇NO₃S
MW: 207,29 g/mol
Melting Pt: 300 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00003835
CAS-Nummer: 103-47-9
EINECS: 203-115-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR