Pipette holders, Clamp-Ette®
Lieferant: Heathrow Scientific
Clamp-Ette®
612-2000EA 173 CHF
612-2000
Pipette holders, Clamp-Ette®
Pipettenständer
Create an instant pipette holding point within the lab.
  • Chemical-resistant polypropylene
  • Fits most major pipette brands
  • Secures clamping for tabletops, benches and shelves
  • Autoclavable
The Clamp-Ette® pipette holder allows you to create an instant pipette holding point within your lab. The holder secures to tabletops, benches and shelves with a twist.