Methyl bromochlorofluoroacetate 98%

Lieferant: Apollo Scientific
Achtung

PC450260-1G PC450260-5G
APOSPC450260-1GEA 276 CHF
APOSPC450260-1G APOSPC450260-5G
Methyl bromochlorofluoroacetate 98%
Methyl bromochlorofluoroacetate
Formel: C₃H₃BrClFO₂
Lager Temperatur: Raumtemperatur
MDL Number: MFCD28054291
CAS-Nummer: 101385-65-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR