Sodium nonafluoropentanoate
Lieferant: Apollo Scientific
|
Achtung
|
PC408085-1G
PC408085-5G
PC408085-250MG
APOSPC408085-1GEA
201
CHF
APOSPC408085-1G
APOSPC408085-5G
APOSPC408085-250MG
Sodium nonafluoropentanoate
Sodium nonafluoropentanoate
Formel:
C₅F₉O₂·Na Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD01325841 CAS-Nummer: 2706-89-0 |