2-Methyl-6-(trifluoromethyl)nicotinaldehyde oxime
Lieferant: Apollo Scientific
PC300656-5G
APOSPC300656-5GEA
0
CHF
APOSPC300656-5G
2-Methyl-6-(trifluoromethyl)nicotinaldehyde oxime
2-Methyl-6-(trifluoromethyl)nicotinaldehyde oxime
Formel:
C₈H₇F₃N₂O MW: 204,149 g/mol Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD20730951 CAS-Nummer: 1227957-39-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...