Methyl 4'-fluoro-1,1'-biphenyl-4-carboxylate

Lieferant: Apollo Scientific

PC300616-5G
APOSPC300616-5GEA 0 CHF
APOSPC300616-5G
Methyl 4'-fluoro-1,1'-biphenyl-4-carboxylate
Methyl 4'-fluoro-1,1'-biphenyl-4-carboxylate
Formel: C₁₄H₁₁FO₂
MW: 230,234 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD05979321
CAS-Nummer: 80254-87-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR