3,3,3-Trifluoro-2-methylpropanoic acid 97%

Lieferant: Apollo Scientific
Gefahr

PC201243-1G
APOSPC201243-1GEA 216 CHF
APOSPC201243-1G
3,3,3-Trifluoro-2-methylpropanoic acid 97%
3,3,3-Trifluoro-2-methylpropanoic acid
Formel: C₄H₅F₃O₂
MW: 142.077 g/mol
Siedepunkt: 67 °C (5 mm Hg)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD19228247
CAS-Nummer: 381-97-5
UN: 3265
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR