7,8-Difluoroquinoline-3-carboxylic acid 95+%
Lieferant: Apollo Scientific
PC201229-1G
PC201229-250MG
APOSPC201229-1GEA
286
CHF
APOSPC201229-1G
APOSPC201229-250MG
7,8-Difluoroquinoline-3-carboxylic acid 95+%
7,8-Difluoroquinoline-3-carboxylic acid
Formel:
C₁₀H₅F₂NO₂ MW: 209,149 g/mol Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD24483950 CAS-Nummer: 318685-41-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...