1-Bromophenazine 95%

Lieferant: Apollo Scientific
Achtung

OR60265-1G OR60265-250MG OR60265-5G
APOSOR60265-1GEA 237 CHF
APOSOR60265-1G APOSOR60265-250MG APOSOR60265-5G
1-Bromophenazine 95%
1-Bromophenazine
Formel: C₁₂H₇BrN₂
Melting Pt: 132…134 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD21086069
CAS-Nummer: 3331-27-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR