2-Bromo-5-cyanobenzoic acid
Lieferant: Apollo Scientific
|
Achtung
|
OR400316-100MG
OR400316-1G
OR400316-250MG
APOSOR400316-100MGEA
52.2
CHF
APOSOR400316-100MG
APOSOR400316-1G
APOSOR400316-250MG
2-Bromo-5-cyanobenzoic acid
2-Bromo-5-cyanobenzoic acid
Formel:
C₈H₄BrNO₂ Melting Pt: 183…185 °C Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD11847005 CAS-Nummer: 845616-12-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...