2,1,3-Benzoxadiazole-5-carbohydrazide

Lieferant: Apollo Scientific
Achtung

OR3609-1G OR3609-5G
APOSOR3609-1GEA 262 CHF
APOSOR3609-1G APOSOR3609-5G
2,1,3-Benzoxadiazole-5-carbohydrazide
2,1,3-Benzoxadiazole-5-carbohydrazide
Formel: C₇H₆N₄O₂
Lager Temperatur: Kühlschrank
MDL Number: MFCD00276976
CAS-Nummer: 175203-93-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR