6-Amino-1H-indazole-7-carboxylic acid

Lieferant: Apollo Scientific
Achtung

Synonyms: 6-Amino-7-carboxy-1H-indazole

OR13445-1G OR13445-250MG
APOSOR13445-1GEA 621 CHF
APOSOR13445-1G APOSOR13445-250MG
6-Amino-1H-indazole-7-carboxylic acid
6-Amino-1H-indazole-7-carboxylic acid
Formel: C₈H₇N₃O₂
MW: 177,16 g/mol
Melting Pt: 180 °C (dec.)
Lager Temperatur: Kühlschrank
MDL Number: MFCD03856017
CAS-Nummer: 73907-95-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR