Tetrahydropyran-2-ylmethylamine hydrochloride

Lieferant: Apollo Scientific
Achtung

OR110077-1G OR110077-5G
APOSOR110077-1GEA 355 CHF
APOSOR110077-1G APOSOR110077-5G
Tetrahydropyran-2-ylmethylamine hydrochloride
Tetrahydropyran-2-ylmethylamine hydrochloride
Formel: C₆H₁₃NO·ClH
MW: 151,634 g/mol
Melting Pt: 142…145 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD06738971
CAS-Nummer: 683233-12-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR