PTB7-Th (PCE10), Reagenz für organische Photovoltaik (OVP)
Lieferant: Thermo Fisher Scientific
H66975.MC
H66975.MD
H66975.ME
H66975.MCEA
1400
CHF
H66975.MC
H66975.MD
H66975.ME
PTB7-Th (PCE10), Reagenz für organische Photovoltaik (OVP)
PTB7-Th (PCE10)
Formel:
(C₄₉H₅₇FO₂S₆)n |
CAS-Nummer:
1469791-66-9 |
Spezifikation Testergebnisse
Appearance (Color) | Black |
Form | Flake or threadlike |
Bandgap energy | 1,58 ± 0,02 eV |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...