pH sensor with integrated temperature detector, SE 102 AN
Lieferant: Knick
662-2356EA 515 CHF
662-2356
pH sensor with integrated temperature detector, SE 102 AN
Elektroden pH-Elektroden
The SE 102-AN sensor with glass body is a reliable sensor for precise pH measurements in the laboratory and in the field. It has an integrated PT 1000 temperature detector. The electrolyte is refillable.
  • Liquid electrolyte, refillable
  • Ceramic junction
  • Integrated temperature detector
  • pH glass with low impedance
  • Suitable for measurements at low temperatures, low alkali error
Application: Precision measurements, chemical applications, aggressive media and quality control.