Triphenylphosphinoxid
Lieferant: Molekula
MOLEM65375399EA
0
CHF
MOLEM65375399
Triphenylphosphinoxid
Triphenylphosphine oxide
Formel:
C₁₈H₁₅OP MW: 278,29 g/mol Siedepunkt: ∼360 °C (1013 hPa) Melting Pt: 155…158 °C Dichte: 1,2 g/cm³ (20 °C) Flash-Pt: 180 °C |
MDL Number:
MFCD00002080 CAS-Nummer: 791-28-6 EINECS: 212-338-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...