Tetramethylammoniumchlorid

Lieferant: LATOXAN
Gefahr

L9078-100 L9078-500
LATOL9078-100EA 0 CHF
LATOL9078-100 LATOL9078-500
Tetramethylammoniumchlorid
Tetramethylammoniumchlorid
Formel: (CH₃)₄NCl
Melting Pt: > 300 °C
Dichte: 1,169 g/cm³ (20 °C)
Flash-Pt: >100 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00011628
CAS-Nummer: 75-57-0
EINECS: 200-880-8
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR