Tetraethylammoniumchlorid

Lieferant: LATOXAN

Synonyms: Tetrylammonium chloride

L9073-25 L9073-5
LATOL9073-25EA 0 CHF
LATOL9073-25 LATOL9073-5
Tetraethylammoniumchlorid
Tetraethylammoniumchlorid
Formel: C₈H₂₀ClN
MW: 165,71 g/mol
Melting Pt: 285 °C
Dichte: 1.08 g/cm³ (20 °C)
MDL Number: MFCD00011828
CAS-Nummer: 56-34-8
EINECS: 200-267-5
Merck Index: 12,09341

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR