Dodecyltrimethylammoniumchlorid
Lieferant: Molekula
|
Achtung
|
MOLE87660455-25GEA
0
CHF
MOLE87660455-25G
Dodecyltrimethylammoniumchlorid
Dodecyltrimethylammonium chloride
Formel:
C₁₅H₃₄ClN MW: 263,89 g/mol Melting Pt: 248…249,5 °C |
MDL Number:
MFCD00041974 CAS-Nummer: 112-00-5 EINECS: 203-927-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...