Dodecyltrimethylammoniumchlorid

Lieferant: Molekula
Achtung

MOLEM11977012EA 0 CHF
MOLEM11977012
Dodecyltrimethylammoniumchlorid
Dodecyltrimethylammonium chloride
Formel: C₁₅H₃₄ClN
MW: 263,89 g/mol
Melting Pt: 248…249,5 °C
MDL Number: MFCD00041974
CAS-Nummer: 112-00-5
EINECS: 203-927-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR